Learn More
ATCC Thermoactinomyces vulgaris
Catalog No. 502379955
Encompass
Thermoactinomyces vulgaris strain NCIB 11364 is a thermophilic bacterium with applications in biotechnology. This strain grows aerobically at 50-55°C.
Catalog No. 50-237-9955 Supplier ATCC Supplier No. 43649
Thermoactinomyces vulgaris strain NCIB 11364 is a thermophilic bacterium with applications in biotechnology. This strain grows aerobically at 50-55°C.