Learn More
ATCC Mycoplasma hyorhinis
Supplier:  ATCC 29052
Encompass
ATCC CCL-10™ (BHK-21) cell culture or a specially formulated cell-free medium* is suitable for propagation and maintenance. *The following publication proposes a medium for the cultivation of this strain, but it has not yet been tested in our lab. Applied and Environmental Microbiology, Vol. 61(5), May l995, p.1976-1979.
Catalog No. 50-238-0836