Learn More
ATCC Escherichia coli
Supplier:  ATCC 11303
Encompass
Escherichia coli strain B [CIP 103914, NCIB 11595, NRC 745] is a whole-genome sequenced bacterium provided in an economical preceptrol format. This product has applications in testing aerosol containment on cell sorters and it is known to produce aspartate aminotransferase.
Catalog No. 50-238-1709