Learn More
ATCC Anaerostipes sp.; Strain 5_1_63FAA
Catalog No. 502380839
Encompass
Anaerostipes sp. strain 5_1_63FAA is an anaerobic bacterium that was isolated from a human subject in Canada as part of the Human Microbiome Project.
Catalog No. 50-238-0839 Supplier ATCC Supplier No. BAA1912
Anaerostipes sp. strain 5_1_63FAA is an anaerobic bacterium that was isolated from a human subject in Canada as part of the Human Microbiome Project.