Learn More
ATCC Escherichia coli
Supplier:  ATCC 10536
Encompass
Escherichia coli strain MacLeod is a whole-genome sequenced bacterial strain that has applications in bactericide testing, sterility testing, and quality control.
Catalog No. 50-238-2302