Learn More
ATCC Escherichia coli
Supplier:  ATCC BAA196
Encompass
Escherichia coli strain J53 pMG223 is a drug-resistant bacterium that was derived from the clinical isolate Klebsiella pneumoniae (ATCC 14714). It produces extended spectrum beta-lactamase (ESBL) TEM-10 and has applications for antimicrobial resistance research and drug development.
Catalog No. 50-238-3680
 Restricted Item