Learn More
ATCC Escherichia coli
Supplier:  ATCC 10798
Encompass
Escherichia coli strain K-12 was isolated from the feces of a diphtheria convalescent. This whole-genome sequenced strain carries F1 and phage lambda (F+ lambda+) and is known to be lytic on Escherichia coli strain W1485 (ATCC 12435).
Catalog No. 50-238-3148