Learn More
ATCC Escherichia coli
Supplier:  ATCC 11775
Encompass
Escherichia coli strain NCTC 9001 is a bacterial type strain that was isolated from urine. This whole-genome sequenced strain can be used for aerosol detection, media testing, and quality control testing.
Catalog No. 50-238-3454