Learn More
ATCC Escherichia coli
Supplier:  ATCC 25404
Encompass
Escherichia coli strain K-12 is a host for transducing phage P1 (ATCC 25404-B1). This product is a whole-genome sequenced strain and is nonlysogenic.
Catalog No. 50-238-4695