Learn More
ATCC Escherichia coli
Supplier:  ATCC 9637
Encompass
This whole-genome sequenced strain of Escherichia coli can be used in susceptibility testing and pharmaceutical testing. This strain is known to produce cephalosporins and 6-aminopenicillanic acid.
Catalog No. 50-238-4827