Learn More
ATCC Escherichia coli; 6 x 0.3mL
Catalog No. 502381765
Encompass
ATCC 15597-MINI-PACK™ consists of 6 ready-to-use vials of ATCC 15597™ frozen in 200 μL of glycerol stock, eliminating the need to rehydrate and culture the strain prior to use. Each vial is provided with a 2-D barcode for easy storage and tracking, as well as peel-off labels for fast and reliable recordkeeping.
Catalog No. 50-238-1765 Supplier ATCC Supplier No. 15597MINIPACK
ATCC 15597-MINI-PACK™ consists of 6 ready-to-use vials of ATCC 15597™ frozen in 200 μL of glycerol stock, eliminating the need to rehydrate and culture the strain prior to use. Each vial is provided with a 2-D barcode for easy storage and tracking, as well as peel-off labels for fast and reliable recordkeeping.