Learn More
ATCC Escherichia coli (Migula) Castellani and Chalmers
Supplier:  ATCC 8739/DEL
Encompass
Escherichia coli strain Crooks is a whole-genome sequenced bacterial strain that was isolated from feces. This product has applications as a quality control strain in testing antimicrobial handwashing formulations, media testing, efficacy testing, and bioresistance testing.
Catalog No. 50-189-128FP