Learn More
ATCC Escherichia coli
Supplier:  ATCC CRM8739
Encompass
Escherichia coli strain Crooks was isolated from feces. This certified reference material can be used in testing and calibration in ISO 17025-accredited laboratories to challenge assay performance, validate or compare test methods, and establish sensitivity, linearity, and specificity during assay validation or implementation.
Catalog No. 50-238-3199