Learn More
ATCC Escherichia coli; Bacteriophage PR772
Catalog No. 502385023
Encompass
This is a PR4-like bacteriophage. It is sensitive to organic solvents and adsorbs to tips of pili coded for by IncP-1, N, and W plasmids.
Catalog No. 50-238-5023 Supplier ATCC Supplier No. BAA769B1
This is a PR4-like bacteriophage. It is sensitive to organic solvents and adsorbs to tips of pili coded for by IncP-1, N, and W plasmids.