Learn More
ATCC Escherichia coli; Bacteriophage T3
Catalog No. 502384127
Encompass
Escherichia coli bacteriophage T3 is a virus propagated in Escherichia coli strain B (ATCC 11303).
Catalog No. 50-238-4127 Supplier ATCC Supplier No. 11303B3
Escherichia coli bacteriophage T3 is a virus propagated in Escherichia coli strain B (ATCC 11303).