Learn More
ATCC Escherichia coli; Bacteriophage T1
Catalog No. 502383108
Encompass
Escherichia coli bacteriophage T1 is a virus that infects and replicates in Escherichia coli strain B (ATCC 11303). This product has been cited in various studies for use as a surrogate for mammalian viruses to test the efficacy of antiviral strategies such as air filtration, sanitizing agents, and ultraviolet germicidal irradiation.
Catalog No. 50-238-3108 Supplier ATCC Supplier No. 11303B1
Add to Cart
Add to Cart
Escherichia coli bacteriophage T1 is a virus that infects and replicates in Escherichia coli strain B (ATCC 11303). This product has been cited in various studies for use as a surrogate for mammalian viruses to test the efficacy of antiviral strategies such as air filtration, sanitizing agents, and ultraviolet germicidal irradiation.