Learn More
ATCC Escherichia coli; bacteriophage T2
Catalog No. 502383467
Encompass
Escherichia coli bacteriophage T2 is a virus that infects and replicates in Escherichia coli strain B (ATCC 11303). This product is provided in an economical preceptrol format.
Catalog No. 50-238-3467 Supplier ATCC Supplier No. 11303B2
 Restricted Item
Add to Cart
Add to Cart
Escherichia coli bacteriophage T2 is a virus that infects and replicates in Escherichia coli strain B (ATCC 11303). This product is provided in an economical preceptrol format.