Learn More
ATCC Escherichia coli; BL21
Catalog No. 502384607
Encompass
Escherichia coli strain BL21 is a whole-genome sequenced bacterium that can be used as a host for bacteriophages T3 (ATCC BAA-1025-B1) and T7 (ATCC BAA-1025-B2).
Catalog No. 50-238-4607 Supplier ATCC Supplier No. BAA1025
Escherichia coli strain BL21 is a whole-genome sequenced bacterium that can be used as a host for bacteriophages T3 (ATCC BAA-1025-B1) and T7 (ATCC BAA-1025-B2).