Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microbiologics™ ESCHERICHIA COLI DERIVED FROM NCTC 13353
SDP

Catalog No. 23047312 Shop All Microbiologics Products
Change view
Click to view available options
Quantity:
2 KWIK-STIK
6 KWIK-STIKS
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-047-312 6 KWIK-STIKS
23-047-310 2 KWIK-STIK
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-047-312 Supplier Microbiologics™ Supplier No. 01265K
Add to Cart
Add to Cart
This item is not returnable. View return policy

Simple, all-inclusive design making life easier for lab techs while reducing chances for errors. Each KWIK-STIK contains a qualitative lyophilized microorganism pellet, ampoule of hydrating fluid and inoculating swab.

  • Self-contained units of a single organism.
  • Available in a pack of 2 or 6, KWIK-STIKs are universally used for all types of microbiological control testing.

Specifications

ATCC® NCTC 13353
Biosafety Level 2
Certifications/Compliance Clinical Laboratory Standards Institute (CLSI)
Content And Storage 2°C to 8°C
Format KWIK-STIK 6 Pack
Manufacturing Origin United States
Product Type Quality Control Organism
For Use With (Application) Antimicrobial Susceptibility Testing, Microbial Identification
Packaging Type Plastic Canister
Quantity 6 KWIK-STIKS
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.