Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microbiologics™ ESCHERICHIA COLI DERIVED FROM NCTC 13353 LYFO DISK
SDP

Catalog No. 23047314 Shop All Microbiologics Products
Change view
Click to view available options
Quantity:
6 Pellets
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-047-314 6 Pellets
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-047-314 Supplier Microbiologics™ Supplier No. 01265L
Add to Cart
Add to Cart
This item is not returnable. View return policy

LYFO DISK, packaged in a glass vial containing 6 pellets of a single strain, offers a ton of flexibility. Simply rehydrate LYFO DISK qualitative microorganism pellets with a diluent, such as phosphate buffer or saline, and you’re ready to inoculate your culture media.

  • 6 lyophilized pellets of a single microorganism strain for quality control

Specifications

ATCC® NCTC 13353
Biosafety Level 2
Certifications/Compliance Clinical Laboratory Standards Institute (CLSI)
Content And Storage 2°C to 8°C
Format LYFO DISK
Manufacturing Origin United States
Product Type Lyfo-Disk QC Control
For Use With (Application) Antimicrobial Susceptibility Testing, Microbial Identification
Packaging Type Glass Vial
Quantity 6 Pellets
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.