Learn More
ATCC Escherichia coli; H10407; serotype O78:H11
Supplier:  ATCC 35401
Encompass
Escherichia coli strain H10407 was isolated from feces and can be used in the detection of LT toxin and Vibrio cholerae ctx A11. This strain produces heat-labile and heat-stable enterotoxins.
Catalog No. 50-238-3020
Edge