Learn More
ATCC Escherichia coli; HB101
Catalog No. 502383953
Encompass
Escherichia coli strain HB101 is an E. coli K12 x E. coli B derivative used for plasmid production.
Catalog No. 50-238-3953 Supplier ATCC Supplier No. 33694
Escherichia coli strain HB101 is an E. coli K12 x E. coli B derivative used for plasmid production.