Learn More
ATCC Escherichia coli; K-12; genomic DNA
Supplier:  ATCC 10798D5
Encompass
Genomic DNA isolated from Escherichia coli strain K-12 (ATCC 10798). This whole-genome sequenced product can be used in PCR and other molecular biology applications.
Catalog No. 50-238-4974