Learn More
ATCC Genomic DNA from Escherichia coli strain MG1655 (ATCC 700926D-5™)
Catalog No. 502383331
Encompass
Genomic DNA from Escherichia coli strain MG1655 (ATCC 700926D-5™) is a high-molecular-weight DNA preparation suitable for PCR and molecular biology applications. Verified by sequencing and electrophoresis, it is stored dried in TE buffer.
Recommended companion product:
Quantitative genomic DNA (Fisher Cat. No. 50-238-5068, ATCC 700926DQ™).
Catalog No. 50-238-3331 Supplier ATCC Supplier No. 700926D5
Genomic DNA from Escherichia coli strain MG1655 (ATCC 700926D-5™) is a high-molecular-weight DNA preparation suitable for PCR and molecular biology applications. Verified by sequencing and electrophoresis, it is stored dried in TE buffer.
Recommended companion product:
Quantitative genomic DNA (Fisher Cat. No. 50-238-5068, ATCC 700926DQ™).