Learn More
ATCC Escherichia coli; MG1655
Catalog No. 502383578
Encompass
Escherichia coli strain MG165 is a host strain with well-characterized restriction maps for NotI, SfiI, XbaI, and BlnI. This bacterial strain can be used as a molecular size marker for pulsed-field gel electrophoresis.
Catalog No. 50-238-3578 Supplier ATCC Supplier No. 47076
Escherichia coli strain MG165 is a host strain with well-characterized restriction maps for NotI, SfiI, XbaI, and BlnI. This bacterial strain can be used as a molecular size marker for pulsed-field gel electrophoresis.