Learn More
ATCC Enterococcus faecalis
Supplier:  ATCC BAA2573
Encompass
Enterococcus faecalis strain [bMx 0502240] is a vancomycin-resistant bacterial strain with applications in antimicrobial resistance research and quality control. The presence of vanA gene was confirmed by PCR.
Catalog No. 50-238-1347