Learn More
ATCC Escherichia coli; O26:H11-Nanoluc
Supplier:  ATCC BAA2580PACK
Encompass
A kit containing reporter-labeled Escherichia coli designed to distinguish control strain cross-contamination from true contamination in test samples. This product is appropriate for use as a positive control in quality control assays for E. coli.
Catalog No. 50-238-4789