Learn More
ATCC Escherichia coli phage; lambda standard
Supplier:  ATCC 23724B2
Encompass
Escherichia coli bacteriophage lambda standard is a bacterial virus that is propagated in Escherichia coli strain C600.
Catalog No. 50-238-2367