Learn More
ATCC Escherichia coli; phage MS2
Catalog No. 502381922
Encompass
Escherichia coli bacteriophage MS2 is a virus that is propagated in Escherichia coli strain C-3000 (ATCC 15597). This bacteriophage has applications in testing water for the presence of coliphages and as a process control for molecular diagnostics.
Catalog No. 50-238-1922 Supplier ATCC Supplier No. 15597B1
Add to Cart
Add to Cart
Escherichia coli bacteriophage MS2 is a virus that is propagated in Escherichia coli strain C-3000 (ATCC 15597). This bacteriophage has applications in testing water for the presence of coliphages and as a process control for molecular diagnostics.