Learn More
ATCC Escherichia coli bacteriophage Phi X174 (ATCC 13706-B1™)
Catalog No. 502383968
Encompass
Escherichia coli bacteriophage Phi X174 (ATCC 13706-B1™) is a somatic coliphage propagated in E. coli strain C. This product has applications in water testing, polyethylene film testing, and bacteriophage detection assays.
Recommended companion product:
Escherichia coli (Fisher Cat. No. 50-238-2221, ATCC 13706™).
Catalog No. 50-238-3968 Supplier ATCC Supplier No. 13706B1
 Restricted Item
Add to Cart
Add to Cart
Escherichia coli bacteriophage Phi X174 (ATCC 13706-B1™) is a somatic coliphage propagated in E. coli strain C. This product has applications in water testing, polyethylene film testing, and bacteriophage detection assays.
Recommended companion product:
Escherichia coli (Fisher Cat. No. 50-238-2221, ATCC 13706™).