Learn More
ATCC Escherichia coli; phage T4
Catalog No. 502381987
Encompass
Due to an upcoming ERP system upgrade, ATCC will have a temporary system shutdown from 11/20/26-12/1/26. All orders will experience a 5-6 days delay in processing. Orders will continue to be sent to ATCC during the blackout and will be held until 12/2/26 when they are released. Any questions, please reach out to Encompass.Supplierchannel@thermofisher.com.
Escherichia coli bacteriophage T4 is a virus that infects and replicates in Escherichia coli strain B. This product is provided in an economical preceptrol format. It has applications for testing aerosol containment on cell sorters and has been cited in various studies for evaluating the antiviral efficacy of copper-binding peptides and intermetallic nanoparticles.
Catalog No. 50-238-1987 Supplier ATCC Supplier No. 11303B4
Due to an upcoming ERP system upgrade, ATCC will have a temporary system shutdown from 11/20/26-12/1/26. All orders will experience a 5-6 days delay in processing. Orders will continue to be sent to ATCC during the blackout and will be held until 12/2/26 when they are released. Any questions, please reach out to Encompass.Supplierchannel@thermofisher.com.
Escherichia coli bacteriophage T4 is a virus that infects and replicates in Escherichia coli strain B. This product is provided in an economical preceptrol format. It has applications for testing aerosol containment on cell sorters and has been cited in various studies for evaluating the antiviral efficacy of copper-binding peptides and intermetallic nanoparticles.