Learn More
ATCC Escherichia coli; phage T5
Supplier:  ATCC 11303B5
Encompass
Escherichia coli bacteriophage T5 is a virus that infects and replicates in Escherichia coli strain B (ATCC 11303). This product has been cited in research evaluating coliphage diversity and activity.
Catalog No. 50-238-4116
 Restricted Item