Learn More
American Research Products Inc Escherichia coli protein E.co
Catalog No. 5000326078
Encompass
Escherichia coli protein (E coli P) ELISA kit from CUSABIO Detection Range 0 156 ng/mL-10 ng/mL Sensitivity 0 039 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26078 Supplier American Research Products Inc Supplier No. CSBE11306O
Escherichia coli protein (E coli P) ELISA kit from CUSABIO Detection Range 0 156 ng/mL-10 ng/mL Sensitivity 0 039 ng/mL Sample serum plasma tissue homogenates Method Sandwich