Learn More
ATCC Escherichia coli; Quantitative DNA
Supplier:  ATCC BAA2440DQ
Encompass
Quantitative Genomic DNA from Escherichia coli strain O111 (ATCC BAA-2440) can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine bacterial load.
Catalog No. 50-238-3143