Learn More
ATCC Escherichia coli; Quantitative DNA
Catalog No. 502384317
Encompass
ATCC Genuine Nucleics can be used for assay development, verification, validation, monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine bacterial load and for other molecular biology applications.
Catalog No. 50-238-4317 Supplier ATCC Supplier No. 43895DQ
 Restricted Item
Add to Cart
Add to Cart
ATCC Genuine Nucleics can be used for assay development, verification, validation, monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine bacterial load and for other molecular biology applications.