Learn More
ATCC Escherichia coli; Strain CDC EDL 1284
Supplier:  ATCC 43893
Encompass
Escherichia coli strain CDC EDL 1284 [929-78] was isolated from human feces. This strain is serotype O124:NM and contains a high-molecular-weight plasmid responsible for enteroinvasiveness.
Catalog No. 50-238-0931