Learn More
ATCC Escherichia coli; Strain 1001728
Catalog No. 502384859
Encompass
Escherichia coli strain 1001728 was isolated from human urine. This strain is resistant to ertapenem and imipenem and was confirmed by PCR to contain the blaNDM gene. The bacterial culture can be used for UTI assay development, antimicrobial resistance research, drug development, and infectious disease research.
Catalog No. 50-238-4859 Supplier ATCC Supplier No. BAA2469
Add to Cart
Add to Cart
Escherichia coli strain 1001728 was isolated from human urine. This strain is resistant to ertapenem and imipenem and was confirmed by PCR to contain the blaNDM gene. The bacterial culture can be used for UTI assay development, antimicrobial resistance research, drug development, and infectious disease research.