Learn More
ATCC Escherichia coli; Strain 1100101
Catalog No. 502384277
Encompass
Escherichia coli strain 1100101 is a whole-genome sequenced bacterium that was isolated from a respiratory sample. This strain is an NDM-1 reference strain and is resistant to ertapenem and imipenem. The bacterium strain can be used in antimicrobial resistance research, drug development, and respiratory disease research.
Catalog No. 50-238-4277 Supplier ATCC Supplier No. BAA2471
Escherichia coli strain 1100101 is a whole-genome sequenced bacterium that was isolated from a respiratory sample. This strain is an NDM-1 reference strain and is resistant to ertapenem and imipenem. The bacterium strain can be used in antimicrobial resistance research, drug development, and respiratory disease research.