Learn More
ATCC Klebsiella pneumoniae; Strain 1101200
Catalog No. 502379655
Encompass
KPC Reference Strain. Resistant to Imipenem and Ertapenem.
Catalog No. 50-237-9655 Supplier ATCC Supplier No. BAA2344
KPC Reference Strain. Resistant to Imipenem and Ertapenem.