Learn More
ATCC Escherichia coli; Strain 1101362
Catalog No. 502382115
Encompass
Escherichia coli strain 1101362 is a drug-resistant bacterium that was confirmed by PCR to contain the blaKPC gene. This strain demonstrates resistance to imipenem and ertapenem and it can be used in antimicrobial resistance research and multidrug testing.
Catalog No. 50-238-2115 Supplier ATCC Supplier No. BAA2340
Escherichia coli strain 1101362 is a drug-resistant bacterium that was confirmed by PCR to contain the blaKPC gene. This strain demonstrates resistance to imipenem and ertapenem and it can be used in antimicrobial resistance research and multidrug testing.