Learn More
ATCC Escherichia coli; Strain 1109131
Supplier:  ATCC BAA2523
Encompass
Escherichia coli strain bMx# 1109131 is a clinical isolate from BioMerieux that can be used as a quality control strain for chromID Oxa 48 medium. This strain is resistant to ertapenem and the depositor claims that this strain is a carbapenemase producer (OXA-48).
Catalog No. 50-238-2543
 Restricted Item