Learn More
ATCC Escherichia coli; Strain 2000-3039
Supplier:  ATCC BAA2193
Encompass
Escherichia coli strain 2000-3039 is a whole-genome sequenced strain that was isolated from stool in Maine. It is toxigenic and was confirmed by PCR to carry the eae and stx1 genes. It has applications in food testing, enteric disease research, and infectious disease research.
Catalog No. 50-238-2866
 Restricted Item