Learn More
ATCC Escherichia coli; Strain 2002-3211
Supplier:  ATCC BAA2219
Encompass
Escherichia coli strain 2002-3211 is a whole-genome sequenced strain that was isolated in Virginia from stool. It is toxigenic and is positive for the eae and stx2 genes and negative for the stx1 gene. This strain is serotype O121:H19 and has applications in enteric disease research and food testing.
Catalog No. 50-238-4805