Learn More
ATCC Escherichia coli; Strain 2003-3014
Catalog No. 502384416
Encompass
Escherichia coli strain 2003-3014 is a whole-genome sequenced strain that was isolated from stool in Michigan. It is toxigenic and was confirmed by PCR to carry the stx1, stx2, and eae genes. This strain is serotype O26:H11 and it has applications for food testing, enteric disease research, and infectious disease research.
Catalog No. 50-238-4416 Supplier ATCC Supplier No. BAA2196
Escherichia coli strain 2003-3014 is a whole-genome sequenced strain that was isolated from stool in Michigan. It is toxigenic and was confirmed by PCR to carry the stx1, stx2, and eae genes. This strain is serotype O26:H11 and it has applications for food testing, enteric disease research, and infectious disease research.