Learn More
ATCC Escherichia coli; Strain 98-3306
Catalog No. 502384414
Encompass
Escherichia coli strain 98-3306 is a whole-genome sequenced strain isolated in South Africa. This strain is serotype O121 and was confirmed by PCR to be negative for the eae, stx1, and stx2 genes.
Catalog No. 50-238-4414 Supplier ATCC Supplier No. BAA2190
Escherichia coli strain 98-3306 is a whole-genome sequenced strain isolated in South Africa. This strain is serotype O121 and was confirmed by PCR to be negative for the eae, stx1, and stx2 genes.