Learn More
ATCC Escherichia coli; Strain 99-3071
Supplier:  ATCC BAA2216
Encompass
Escherichia coli strain 99-3071 is a whole-genome sequenced strain that was isolated from stool. It is non-toxigenic and is a negative control for STEC virulence genes. This strain is serotype O145:H34.
Catalog No. 50-238-3094