Learn More
ATCC Escherichia coli; Strain CN13
Catalog No. 502384987
Encompass
Bacteriophage host. Nalidixic acid-resistant mutant of ATCC 13706.
Catalog No. 50-238-4987 Supplier ATCC Supplier No. 700609
Bacteriophage host. Nalidixic acid-resistant mutant of ATCC 13706.