Learn More
ATCC Escherichia coli
Catalog No. 502380496
Encompass
Escherichia coli strain J53 pMG224 is a multidrug-resistant bacterium that produces extended spectrum beta-lactamase (ESBL) TEM-12. This strain is whole-genome sequenced and has applications in antimicrobial resistance research.
Catalog No. 50-238-0496 Supplier ATCC Supplier No. BAA197
Escherichia coli strain J53 pMG224 is a multidrug-resistant bacterium that produces extended spectrum beta-lactamase (ESBL) TEM-12. This strain is whole-genome sequenced and has applications in antimicrobial resistance research.