Learn More
Sigma Aldrich Fine Chemicals Biosciences Escherichia coli Strain K1
Catalog No. 502904825
Encompass
Escherichia coli (E.coli) is a non-spore forming Gram-negative rod-shaped facultative anerobe which is found in the human gastrointestinal tract. It belongs to the family of Enterobacteriaceae. E.coli is chemoorganotrophic and grows at 37C. Pathogenic E.coli strains is associated with diarrhea septicemia meningitis and urinary tract infections. It inhibits the colonization of the gut by harmful bacteria. E.coli acts as an indicator of fecal contamination.
Catalog No. 50-290-4825 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. EC15G
Escherichia coli (E.coli) is a non-spore forming Gram-negative rod-shaped facultative anerobe which is found in the human gastrointestinal tract. It belongs to the family of Enterobacteriaceae. E.coli is chemoorganotrophic and grows at 37C. Pathogenic E.coli strains is associated with diarrhea septicemia meningitis and urinary tract infections. It inhibits the colonization of the gut by harmful bacteria. E.coli acts as an indicator of fecal contamination.