Learn More
ATCC Escherichia coli; Strain P68
Catalog No. 502383671
Encompass
Escherichia coli strain P68 is a whole-genome sequenced bacterium that was isolated from cattle hides. This strain is non-toxigenic and is recommended by the USDA as a surrogate indicator organism to measure changes in microbial counts.
Catalog No. 50-238-3671 Supplier ATCC Supplier No. BAA1431
Escherichia coli strain P68 is a whole-genome sequenced bacterium that was isolated from cattle hides. This strain is non-toxigenic and is recommended by the USDA as a surrogate indicator organism to measure changes in microbial counts.